Anti-LRRN4 Rabbit Polyclonal Antibody
PRSI71-079
- Antigen name:Leucine Rich Repeat Neuronal 4
- Antigen symbol:LRRN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Human
- Concentration:1 mg/mL
- Immunogen:Synthetic 17 amino acid peptide from C-Terminus of human LRRN4 / C20orf75.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
LRRN4 / C20orf75 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 2.5 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LRRN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human