Anti-ITGA2 Rabbit Polyclonal Antibody
PRSI48-404
- Antigen name:Integrin Alpha 2
- Antigen symbol:ITGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gibbon,Human
- Form:Liquid
- Antigen synonyms:BR|GPIa|VLAA2|DX5 antigen|pan-NK cells|CD49B|HPA-5|VLA-2|ITGA2
- Immunogen:Integrin alpha 2 antibody was raised against a peptide located near the Extracellular domain of Integrin alpha 2 (Human).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
ITGA2 encodes integrin alpha chain 2. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. The I-domain containing alpha integrin 2 combines with beta integrin 1 to form a collagen-binding integrin referred to as glycoprotein Ia/IIa when expressed on platelets, and very late (activation) antigen 2 ('VLA-2') when found on T-cells. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ITGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human