Anti-PROK2 Goat Polyclonal Antibody
PRSI42-268
- Antigen name:Prokineticin 2
- Antigen symbol:PROK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:BV8|MIT1|KAL4|PK2|HH4
- Concentration:500 ug/mL
- Immunogen:Prokineticin 2 antibody was raised against a 15 amino acid synthetic peptide near the internal region of Prokineticin 2.
- Size:100 µg
- Pk:100 µG
Prokineticin 2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Prokineticin 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PROK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0