Anti-CDK4 Rabbit Polyclonal Antibody
Catalog # PRSI30-994
Supplier: ProSci Inc.
Specifications
- Antigen name:Cyclin-dependent Kinase 4
- Antigen symbol:CDK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CMM3|MGC14458|PSK-J3|Melanoma Cutaneous Malignant 3|Cell Division Kinase 4|Cell Division Protein Kinase 4
- Immunogen:A synthetic peptide corresponding a region of human CDK4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Cdk4 is probably involved in the control of the cell cycle.
Antibody is purified by Protein A/G affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CDK4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: