Anti-SLC14A1 Rabbit Polyclonal Antibody
Catalog # PRSI30-354
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 14 Member A1
- Antigen symbol:SLC14A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:UTE|UT1|RACH2|JK|HUT11|HsT1341|UT-B1|RACH1
- Immunogen:A synthetic peptide corresponding a region of human SLC14A1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC14A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human