Anti-SEC14L2 Rabbit Polyclonal Antibody
Catalog # PRSI27-406
Supplier: ProSci Inc.
Specifications
- Antigen name:SEC14-like 2
- Antigen symbol:SEC14L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:C22orf6|TAP1|SPF|TAP
- Immunogen:A synthetic peptide corresponding a region of human SEC14L2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SEC14L2 encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SEC14L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human