Order Entry
Germany
ContactUsLinkComponent
Anti-RNASET2 Rabbit Polyclonal Antibody
Anti-RNASET2 Rabbit Polyclonal Antibody
Catalog # PRSI25-117
Supplier:  ProSci Inc.
undefined
Anti-RNASET2 Rabbit Polyclonal Antibody
Catalog # PRSI25-117
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Ribonuclease T2
  • Antigen symbol:
    RNASET2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    RNASET2|RNASE6PL|bA514O12.3|Ribonuclease 6
  • Immunogen:
    A synthetic peptide corresponding a region of human RNASET2.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

This ribonuclease gene is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human malignancies and chromosomal rearrangement.This ribonuclease gene is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human malignancies and chromosomal rearrangement. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.

RNASET2 antibody can be used for detection of RNASET2 by ELISA at 1:62500. RNASET2 antibody can be used for detection of RNASET2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RNASET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human