Specifications
- Antigen name:Serine/threonine Kinase 11
- Antigen symbol:STK11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:PJS|Renal carcinoma antigen NY-REN-19|hLKB1|LKB1|STK11|Liver kinase B1|Serine/threonine-protein kinase STK11
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:LKB1 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human LKB1.
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
LKB1 Antibody: The LKB1 serine/threonine protein kinase was initially identified as a tumor suppressor gene mutated in human Peutz-Jeghers syndrome (PJS), a condition resulting in the growth of numerous intestinal polyps classed as hamartomas. LKB1 exists as a heterotrimeric complex with two other proteins, Ste20-related adaptor protein (STRAD) and MO25. Together, this complex can phsophorylate and activate the AMP-activate protein kinase (AMPK). Following AMPK activation by LKB1, AMPK then phosphorylates TSC1 and TSC2, key components of the metabolism-regulating TOR signaling pathway, which antagonizes the activation for the TOR pathway. LKB1 has also been shown to play a fundamental role in controlling the spatial orientation of structures required to maintain an ordered, polarized epithelium.
Type:
Antigen: STK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse