Orders LinkContactUsLinkComponent
Drip pads
Drip pads
Drip pads
Certificates
Drip pads
4 Options Available Below
Product Details & Documents
Drip pads can capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable
  • Made from UV and chemical-resistant modified PVC
  • Coated mesh outer layer to trap coarse debris
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs.
Certifications: Helps to comply with EPA for spill prevention (SPCC).
Product Family Options
Product Information
  • Capacity
    L×W
    Availability
    Price
  • 119-0352
    Pad drip extended refills
    Pad drip extended refills
    5 l
    483 × 381 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 119-0348
    Pad drip standard
    Pad drip standard
    5 l
    584 × 533 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 194,00 €
    194,00 €
    1 * 1 ST
       194,00 €
  • 119-0349
    Pad drip extended
    Pad drip extended
    7 l
    914 × 610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • 119-0353
    Pad drip refills
    Pad drip refills
    7 l
    1321 × 762 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,90 €
    87,90 €
    1 * 1 ST
       87,90 €