About this item
A robust design made of 55% recycled polyethylene features convenient rolltop doors to allow access from front or rear.
- Eco-friendly, weather-resistant drum storage
- Spacious interior for funnels or pumps
- Forklift pockets front and back for easy transport
- Ships fully assembled
Weather-resistant outdoor sheds offer high performance spill control. Forklift pockets on the front and back enable easy transport and relocations. The leakproof pallet sump features thick, durable deck grating that can be removed for easy inspection and clean up of spills. Wide door opening of 132,1 cm (52") facilitates easy storage palletised loads.
A spacious interior offers ample headspace to accommodate working operations with funnels or pumps. Safety features include grounding capability and integral anchor bolt locations for wind resistance. Molded in lugs accept optional padlocks for security. Ships fully assembled to save on set-up time. Optional ramp simplifies drum placement.