Drain covers, water-filled
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
These water filled drain covers ensure regulatory compliance and prevent run-off pollutants from entering stormwater drains.
- Easy open push plug for filling with standard water hose
- Deploy cover as a preventative measure to seal drains before washdowns, maintenance, or other clean-up tasks that may allow contaminants into nearby water systems
- Long lasting durable modified vinyl construction resists damage caused by UV rays
Certifications: Helps to comply with EPA for stormwater regulations
Ordering information: Order a cover size that overlaps drain opening at least 152 mm on each edge.
Caution: To prevent edges from losing contact with ground surface, do not fill over 127 mm in height.