Boom diverters, water-filled
These water-filled boom diverters temporarily divert and control fluids for easy clean-up or recovery processes. The flexible PVC film construction forms a diversionary barrier also on uneven surfaces such as asphalt or concrete to form a diversionary barrier away from storm drains or as a holding area for wet vacuum operations.
- Easy-open push plug for filling with standard water hose
- Redirects liquids away from work areas and storm drains
Water filled booms are used in storm water management, portable washdown/pressure washing diversion, spill diversion, and equipment and personnel decontamination.
: Helps to comply with EPA for stormwater regulations.