Nitrocefin ≥95% (by UV method) dye, Calbiochem®, Sigma-Aldrich®
484400-5MG
:
- Pack type:Glass bottle
- Pk:5 mg
- Seq:2210
- Purity:≥95% by UV
- Form:Orange-yellow solid
- Storage:-20 °C
:
:516.499 g/mol
:Packaged under inert gas.
:Freezer
:41906-86-9
:C₂₁H₁₆N₄O₈S₂
:MFCD00864896