Order Entry
Germany
Orders LinkContactUsLinkComponent
Prednicarbate, Reference Standards

Prednicarbate, Reference Standards

Supplier:   USP
Prednicarbate, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1554909.
  • 1554910.
  • 1554920.
  • 1554931.
  • Description:
  • Prednicarbate
  • Prednicarbate Related Compound A
  • Prednicarbate Related Compound B
  • Prednicarbate Related Compound C
  • Synonym(s):
  • 1,2-dihydroprednicarbate
  • Prednisolone-17-ethylcarbonate
  • Prednisolone-21-propionate
  • CAS No.:
  • 73771-04-7
  • 671225-26-6
  • 104286-02-4
  • 5740-62-5
  • Formula:
  • C₂₇H₃₆O₈
  • C₂₇H₃₈O₈
  • C₂₄H₃₂O₇
  • C₂₄H₃₂O₆

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1554909.
    Prednicarbate
    Prednicarbate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 348,00 €
    348,00 €
    1 * 200 mg
       348,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1554909.
    Description
    Prednicarbate
    Synonym(s)
    CAS No.
    73771-04-7
    Formula
    C₂₇H₃₆O₈
  • 1554910.
    Prednicarbate Related Compound A
    Prednicarbate Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 330,00 €
    1 330,00 €
    1 * 20 mg
       1 330,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1554910.
    Description
    Prednicarbate Related Compound A
    Synonym(s)
    1,2-dihydroprednicarbate
    CAS No.
    671225-26-6
    Formula
    C₂₇H₃₈O₈
  • 1554920.
    Prednicarbate Related Compound B
    Prednicarbate Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 507,00 €
    507,00 €
    1 * 20 mg
       507,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1554920.
    Description
    Prednicarbate Related Compound B
    Synonym(s)
    Prednisolone-17-ethylcarbonate
    CAS No.
    104286-02-4
    Formula
    C₂₄H₃₂O₇
  • 1554931.
    Prednicarbate Related Compound C
    Prednicarbate Related Compound C
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 507,00 €
    507,00 €
    1 * 20 mg
       507,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1554931.
    Description
    Prednicarbate Related Compound C
    Synonym(s)
    Prednisolone-21-propionate
    CAS No.
    5740-62-5
    Formula
    C₂₄H₃₂O₆
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.