Order Entry
Germany
ContactUsLinkComponent
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Catalog # 2851-100ML
CAS Number:  
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Catalog # 2851-100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Citrate phosphate buffer concentrate
  • M-Clarity quality level:
    MQ500

Specifications

About this item

These buffers are used to elute adult hemoglobin by the procedure of Oski and Naiman, a modification of the Kleihauer Betke procedure for the demonstration of fetal hemoglobin.