Beta Carotene, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1065480.
- 1065491.
- Description:
- Beta Carotene
- Beta Carotene System Suitability
- CAS No.:
- 7235-40-7
- Formula:
- C₄₀H₅₆
Specifications
Product Family Options
Product Information
- StandardsPk InfoPack typePkAvailabilityPrice
- 1065480.Beta CaroteneBeta CaroteneUSP reference standard7 x 100 mgAmpule700 mgLog in for availability1 * 700 mg380,00 €Specifications:
More
Specifications
Cat. No.1065480.DescriptionBeta CaroteneCAS No.7235-40-7FormulaC₄₀H₅₆ - 1065491.Beta Carotene System SuitabilityBeta Carotene System SuitabilityUSP reference standardVial200 mgLog in for availability1 * 200 mg432,00 €Specifications:
More
Specifications
Cat. No.1065491.DescriptionBeta Carotene System SuitabilityCAS No.Formula



