Order Entry
Germany
Orders LinkContactUsLinkComponent
Paclitaxel, Reference Standards

Paclitaxel, Reference Standards

 :   USP
Paclitaxel, Reference Standards
 :  USP
 
Paclitaxel, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1491332.
  • 1491343.
  • 1491354.
  • Description:
  • Paclitaxel
  • Paclitaxel Related Compound A
  • Paclitaxel Related Compound B
  • Synonym(s):
  • Cephalomannine
  • 10-deacetyl-7-epipaclitaxel
  • CAS No.:
  • 33069-62-4
  • 71610-00-9
  • 78454-17-8
  • Formula:
  • C₄₇H₅₁NO₁₄
  • C₄₇H₅₁NO₁₄
  • C₄₅H₄₉NO₁₃

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Paclitaxel
    USP reference standard
    Vial
    200 mg
       
       
    1 * 300 mg
    1 * 300 mg - 3 520,00 €
    3 520,00 €
    1 * 300 mg
       3 520,00 €

     
     
    Specifications
    Cat. No.
    1491332.
    Description
    Paclitaxel
    Synonym(s)
    CAS No.
    33069-62-4
    Formula
    C₄₇H₅₁NO₁₄
  • Paclitaxel Related Compound A
    USP reference standard
    Vial
    20 mg
       
       
    1 * 20 mg
    1 * 20 mg - 1 820,00 €
    1 820,00 €
    1 * 20 mg
       1 820,00 €

     
     
    Specifications
    Cat. No.
    1491343.
    Description
    Paclitaxel Related Compound A
    Synonym(s)
    Cephalomannine
    CAS No.
    71610-00-9
    Formula
    C₄₇H₅₁NO₁₄
  • Paclitaxel Related Compound B
    USP reference standard
    Vial
    20 mg
       
       
    1 * 20 mg
    1 * 20 mg - 2 000,00 €
    2 000,00 €
    1 * 20 mg
       2 000,00 €

     
     
    Specifications
    Cat. No.
    1491354.
    Description
    Paclitaxel Related Compound B
    Synonym(s)
    10-deacetyl-7-epipaclitaxel
    CAS No.
    78454-17-8
    Formula
    C₄₅H₄₉NO₁₃
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.