ContactUsLinkComponent
Lithium ore, spodumene - lithium oxide, NIST standards
Lithium ore, spodumene - lithium oxide, NIST standards
Catalog # PROONIST-182
CAS Number:  
Lithium ore, spodumene - lithium oxide, NIST standards
Catalog # PROONIST-182
CAS Number:  
Specifications
  • Pk:
    45 g
  • Description:
    NIST-182, lithium ore, petalite - lithium oxide
Specifications
About this item
This reference standards are intended to use in checking the accuracy of assay methods. These are supplied in fine powder form.
Certifications: Traceable to NIST and SRM numbers.