Halogenated and substituted benzene and phenol standards, trichlorophenol
PROOCILULM75251.2
:
- Pk:1,2 mL
- CAS No.:95-95-4
- Description:2,4,5-Trichlorophenol (unlabeled), 100 µg/ml in methanol
- Formula:Cl₃C₆H₂OH
- Description:2,4,5-Trichlorophenol (unlabeled)