Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pyrethroid pesticide and metabolite standards, 3-Phenoxybenzoic acid
Pyrethroid pesticide and metabolite standards, 3-Phenoxybenzoic acid
Pyrethroid pesticide and metabolite standards, 3-Phenoxybenzoic acid
Supplier:
LGC Standards PROMOCHEM
Certificates
Expand image
Pyrethroid pesticide and metabolite standards, 3-Phenoxybenzoic acid
Supplier:
LGC Standards PROMOCHEM
About this item
Specifications
CAS No.:
3739-38-6
Formula:
C₁₃H₁₀O₃
Description:
3-Phenoxybenzoic acid
Specifications