Impurity and API standard, sibutramine hydrochloride monohydrate
PROOMM0991.00
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:125494-59-9
- Description:Sibutramine hydrochloride monohydrate
- Formula:C₁₇H₂₉Cl₂NO
- Description:Sibutramine hydrochloride monohydrate