Impurity and API standard, L-amoxicillin
Catalog # PROOMM0232.02
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:26889-93-0
- Description:(2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
- Formula:C₁₆H₁₉N₃O₅S
- Description:(2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Amoxicillin)