Order Entry
Germany
ContactUsLinkComponent
Impurity and API standard, 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride
Impurity and API standard, 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride
Catalog # PROOMM0021.01
CAS Number:  
Impurity and API standard, 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride
Catalog # PROOMM0021.01
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    1006-21-9
  • Description:
    6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol hydrochloride
  • Formula:
    C₈H₉NO₂·ClH
  • Description:
    6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride

Specifications

About this item