Impurity and API standard, 2-(5-nitro-1H-imidazol-1-yl)ethanol
Catalog # PROOMM0012.07
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:5006-68-8
- Description:2-(5-nitro-1H-imidazol-1-yl)ethanol
- Formula:C₅H₇N₃O₃
- Description:2-(5-Nitro-1H-imidazol-1-yl)ethanol