Organic reference standards, Miconazole, BP
Catalog # PROOBP884
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:25 mg
- Standards:Reference standard
- CAS No.:24155-42-8
- Description:Miconazole impurity A
- Formula:C₁₁H₁₀Cl₂N₂O
- Description:Miconazole impurity A