Organic reference standards, Lynestrenol standard, BP
Catalog # PROOBP776
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:25 mg
- Standards:Reference standard
- CAS No.:52-76-6
- Description:Lynestrenol standard
- Formula:C₂₀H₂₈O
- Description:Lynestrenol