Order Entry
Germany
ContactUsLinkComponent
Yeast mating factor alpha (from yeast)
Yeast mating factor alpha (from yeast)
Catalog # GENSRP01002
Supplier:  Genscript
CAS Number:  
Yeast mating factor alpha (from yeast)
Catalog # GENSRP01002
Supplier:  Genscript
CAS Number:  

Specifications

  • Pk:
    10 mg
  • CAS No.:
    59401-28-4
  • Source:
    Yeast
  • Species:
    Yeast
  • Storage conditions:
    –20 °C
  • Protein/peptide name:
    Mating factor alpha
  • Purity:
    <95% (HPLC)
  • Solubility:
    Soluble in water
  • Molecular weight:
    1683,98
  • Sequence:
    WHWLQLKPGQPMY (one-letter code)

    {TRP}{HIS}{TRP}{LEU}{GLN}{LEU}{LYS}{PRO}{GLY}{GLN}{P
    RO}{MET}{TYR}
    (three-letter code)
  • Colour:
    White
  • Concentration:
    5 µM - 100 µM
  • Molecular formula:
    C₈₂H₁₁₄N₂₀O₁₇S

Specifications

About this item

This alpha factor pheromone arrests yeast in the G1 phase of their cell cycle. It helps to induces the expression of mating gene, changes in nuclear architecture and consequently polarizes the growth toward the mating partner.

STE2 encodes the presence of alpha factor pheromone receptor on mating type A cells in yeast.