Orders LinkContactUsLinkComponent
Handheld microscope, BMS Trekker
Handheld microscope, BMS Trekker
Handheld microscope
Catalog # BREU76520
Supplier:   BREUKHOVEN
Handheld microscope, BMS Trekker
Catalog # BREU76520
Supplier:  BREUKHOVEN
Handheld microscope, BMS Trekker
Handheld microscope
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The BMS Trekker is a high quality handheld microscope that enhances objects up to 35 times.
  • Detachable eyepiece to be used as a 10× magnification latch
  • Built-in white LED with daylight correction – efficient energy saving lamp
  • Can be connected to any SLR camera
Suitable for outdoor and indoor use in technology, science, schools and leisure.
Specifications
  • Magnification:
    10× - 35×
  • Description:
    Handheld microscope
  • Cat. No.:
    BREU76520
  • Illumination:
    LED
  • Electrical:
    9 V PP battery
  • Weight:
    215 g