Conductivity and TDS standards, natural water
Supplier: Hach
Certificates
About this item
Natural Water Standard Solution:40% sodium sulphate/40% sodium bicarbonate/20% sodium chloride.
Natural Water Standard Solution:40% sodium sulphate/40% sodium bicarbonate/20% sodium chloride.