0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, loratadines, Ph. Eur.
Organic reference standards, loratadines, Ph. Eur.
:
EDQM
Organic reference standards, loratadines, Ph. Eur.
:
EDQM
CAS No.:
79794-75-5
Formula:
C₂₂H₂₃ClN₂O₂
Description:
Loratadine for system suitability