Cold resistant gloves, Showa 451
Supplier: Showa
Certificates
About this item
These acrylic/cotton/polyester liners with latex coating on palm and fingertips are ideal for heavy duty handlings in cold environments.
- Abrasion resistant and and very pleasant to wear in cold weather conditions
- Protects the hand in damp environments or against aggressive detergents or alcohols
- Breathable back of hand to reduce perspiration
- Surface provides tactile feel and better grip
- Anatomical shape: Designed for easy movement and continuous wear
- No irritation by seams in contact with the skin
- Low-soil colour
Typical applications include: Agriculture, construction, horticulture, packaging, railways, warehousing & distribution.
Certifications: EN 388:2016 (2241X), EN 511 (010); Cat. II
Specifications
- Catalog No:
- 112-5837
- 112-5836
- 112-5834
- 112-5835
- Model:
- Showa 451
- Showa 451
- Showa 451
- Showa 451
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyester/cotton/acrylic
- Polyester/cotton/acrylic
- Polyester/cotton/acrylic
- Polyester/cotton/acrylic
- Finish:
- Rough
- Rough
- Rough
- Rough
- Cuff style:
- Elastic
- Elastic
- Elastic
- Elastic
- EN norm:
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)