Order Entry
Germany
ContactUsLinkComponent
 
Cut resistant gloves, Split Cut
Catalog # DALL2049297/9
Supplier:  HONEYWELL SAFETY
undefined
Cut resistant gloves, Split Cut
Catalog # DALL2049297/9
Supplier:  HONEYWELL SAFETY

Specifications

  • Glove size:
    9
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Grey
  • Length:
    300 mm
  • Seq:
    01
  • Thickness:
    0,9 - 1,1 mm
  • EN norm:
    EN 388:2016 (4X42D)
  • Liner material:
    Dyneema®/polyamide/glass
  • Gauge:
    10
  • Pk:
    1 PAIR

Specifications

About this item

These split cowhide leather gloves are fully lined with a combination of Dyneema®/polyamide/glass. The gloves provide excellent resistance to mechanical hazards such as cuts and abrasion in highly aggressive environments.

  • Seamless knit construction moulds the natural shape of the hand
  • Split leather provides good flexibility
  • Liner provides an exceptional cut (level 5) and abrasion resistance (level 4)
  • Kevlar® yarn strengthens the stitching
  • Gunn cut, winged thumb
  • Latex wrist, vein protection

Ideal for heavy handling with cut hazards in greasy or oily conditions. Typical applications include: Aviation, building and construction, glass industries, green spaces, iron and steel industry, metal steel, paper industries.

Certifications: EN 388:2016 (4×42D); Cat. II