Cut resistant gloves, Aracut® Grip
Supplier: HONEYWELL SAFETY
Certificates
About this item
These knitted Kevlar® gloves feature blue PVC dots on the palm. They offer excellent cutting resistance and protection against thermal hazards in a dry atmosphere.
- Knitting technique helps to make a glove that conforms closely to the hand, with good dexterity
- Optimal comfort with added sturdiness
- PVC dots ensure greater abrasion resistance and a better grip
Ideal for the handling of sharp and possibly hot parts in a dry environment. Typical applications include: Industry, automotive and part manufacturer, building and construction, glass industries, iron and steel industry, metal steel, paper industries.
Certifications: EN 388:2016 (0X4XB); Cat. II
Specifications
- Catalog No:
- DALL2032085/9
- DALL2032085/8
- DALL2032085/7
- DALL2032085/10
- Model:
- Grip
- Grip
- Grip
- Grip
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- PVC dots on palm
- PVC dots on palm
- PVC dots on palm
- PVC dots on palm
- Gauge:
- 10
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (0X4XB)
- EN 388:2016 (0X4XB)
- EN 388:2016 (0X4XB)
- EN 388:2016 (0X4XB)