VWR® B'Wipes 6000E, Cleanroom floor wipes
Catalog # 115-1830
Supplier: VWR Collection
Specifications
- Packed:Stacked, 1×50
- Description:Series 6000E floor wipes
- L×W:250×600 mm
- Seq:12
- Pk:50
Specifications
About this item
Double ply polyester sealed-edge wipe for use in ISO 5 cleanrooms.
- Economical version of the series 6000, especially for floor cleaning
- Two plies quilted for superior absorption
- Ideal for applying a large quantity of cleaning/disinfecting solution on the floors, maximising the number of square metres covered with a single wipe