Order Entry
Germany
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)rCP) [clone: RM4-5]
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)rCP) [clone: RM4-5]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)rCP) [clone: RM4-5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 67-0042-U100
  • 67-0042-U025
  • Antigen symbol:
  • CD4
  • CD4
  • Antigen synonyms:
  • L3T4, T4
  • L3T4, T4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)
  • PerCP (Peridinin-Chlorophyll Protein Complex)
  • Clone:
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications