Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # 22-8067-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Epitope:Heavy and light chain
- Storage temperature:Store undiluted liquid at 2-8°C.
- Pk:1 mg
Specifications
About this item
Goat anti-Rabbit IgG (H&L) - Affinity Pure, Alkaline Phosphatase Conjugate
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit