Order Entry
Germany
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 72-8080-M001
Supplier:  Tonbo Biosciences
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 72-8080-M001
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Goat
  • Western blot:
    Yes
  • Epitope:
    Heavy and light chain
  • Cross adsorption:
    Yes
  • Storage temperature:
    Store freeze-dried powder at 2-8 °C. Once rehydrated and in solution store at 2-8 °C.
  • Pk:
    1 mg

Specifications

About this item

Donkey anti-Goat IgG (H&L) - Affinity Pure, HRP Conjugate, min x w/human, mouse, rabbit or rat IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Goat