Anti-IgM (µ chain) Rabbit Polyclonal Antibody
70-8075-M001
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:Human IgM (µ chain)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:µ chain
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Rabbit anti-Human IgM (mu chain) - Affinity Pure
Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human