Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
35-8066-M002
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Epitope:µ chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Goat anti-Mouse IgM (mu chain) - Affinity Pure, FITC Conjugate, min x w/human IgG or serum proteins
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Mouse