Anti-IgA Goat Polyclonal Antibody (Biotin)
Catalog # 30-8020-U500
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin A alpha
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:α chain
- Antibody fragment:F(ab')2
- Storage temperature:Store at 2-8 °C
- Size:0.5 mg
- Pk:0,5 mg
Specifications
About this item
Goat anti-Human IgA (alpha chain), F(ab)'2 Fragment - Affinity Pure, Biotin Conjugate
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: α chain
Host: Goat
Isotype:
Reactivity: Human