Specimen collection and transport kits, Capitol Vial™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Capitol Vial™ are specimen containers manufactured airtight, leak resistant and aseptic by process.
- Twin valve design which creates a triple seal at the flip cap and vial interface
- Superior leak resistant performance
- Specimen samples are contained during transport and storage
Capitol Vial™ kits are sealed inside the collection cup with a foil top lid for transport (in accordance with IATA requirements) or other applications where a second sample is required.
Delivery: Capitol Vial™ kits contain 1×240 ml beaker with temperature indicator strip; 1× (single foil top) or 2× (split foil top) 45 ml tamper evident vial(s); dual pocket specimen transport bag with absorbent pad; and foil top. The hair/nail specimen collection kit includes foil, envelope and sealing tape.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications
- Catalog No:
- 216-2138
- 216-2139
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement