Order Entry
Germany
ContactUsLinkComponent
Test kits, photometric, lead, LCK cuvette tests
Test kits, photometric, lead, LCK cuvette tests
Catalog # 705-3063
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, lead, LCK cuvette tests
Catalog # 705-3063
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    PAR
  • Pk:
    25 Tests
  • Description:
    Lead
  • Seq:
    237
  • Type:
    LCK306
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,1 - 2,0 mg/l

Specifications

About this item

Ready for use, ensures maximum safety, easy to handle, photometric evaluation system and environmentally friendly.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour coding

Storage conditions: 15 to 25 °C

Ordering information: For Total Lead digestion use CRACK SET LCW902.