Order Entry
Germany
ContactUsLinkComponent
Test kits, photometric, Starch, LCK cuvette tests
Test kits, photometric, Starch, LCK cuvette tests
Catalog # 705-3004
Supplier:  Hach
CAS Number:  
Test kits, photometric, Starch, LCK cuvette tests
Catalog # 705-3004
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Hach Method
  • Pk:
    25 Tests
  • Description:
    Starch
  • Seq:
    268
  • Type:
    LCK357
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    2 - 150 mg/l Starch

Specifications

About this item

Pre-dosed reagents for maximum safety.

Storage conditions: 15 to 25 °C