Order Entry
Germany
ContactUsLinkComponent
Test kits, photometric, Nitrite (trace), LCK cuvette tests
Test kits, photometric, Nitrite (trace), LCK cuvette tests
Catalog # 705-3056
Supplier:  Hach
CAS Number:  
Test kits, photometric, Nitrite (trace), LCK cuvette tests
Catalog # 705-3056
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Diazotisation
  • Pk:
    50 Tests
  • Description:
    Nitrite
  • Seq:
    253
  • Type:
    LCK541
  • For unit:
    DR3900, DR6000, DR2800, DR3800, DR5000
  • Range:
    0,0015 - 0,03 mg/l NO₂-N
    0,005 - 0,1 mg/l NO₂

Specifications

About this item

Pre-dosed reagents for maximum safety.

Storage conditions: 2 to 8 °C

Certifications: EN ISO 26777, DIN 38405 D10