ContactUsLinkComponent
Stitch cutter, disposable, 0526
Stitch cutter, disposable, 0526
Catalog # SWAN0526
Supplier:  SWANN MORTON
undefined
Stitch cutter, disposable, 0526
Catalog # SWAN0526
Supplier:  SWANN MORTON
Specifications
  • Description:
    Stitch cutter, sterile, disposable
  • Pk:
    10
Specifications
About this item
Sterile stitch cutter mounted on a plastic handle allowing extra dexterity and distance from the wound.
  • Stainless steel blade
  • PS handle
Used for pain free removal of modern day suture materials on healing of the wound.