Order Entry
Germany
ContactUsLinkComponent
Urea 98.0-102.0% USP, J.T.Baker®
Urea 98.0-102.0% USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Urea 98.0-102.0% USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets U.S.P Requirements:
    98.0 - 102.0%
  • GMP Manufactured Product:
    ≤0.04%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (NH₂CONH₂):
    98.0 - 102.0 %
  • USP - Alcohol Insoluble Matter:
    ≤ 0.04 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Organic impurities: Urea related compound A:
    ≤ 0.1 %
  • USP - Organic impurities: Any individual unspecified impurity:
    ≤ 0.1 %
  • USP - Organic impurities: Total impurities:
    ≤ 2.0 %
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • Absorbance of a 5M Solution - 260 nm:
    ≤ 0.050
  • Absorbance of a 5M Solution - 280 nm:
    ≤ 0.030
  • Biuret:
    ≤ 0.01
  • Conductivity of 8.5M Solution at 25°C, (µmho/cm):
    ≤ 30
  • Cyanate (CNO):
    None Detected
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • Insoluble Matter:
    ≤ 0.01 %
  • Water (H₂O):
    ≤ 0.5 %
  • Chloride (Cl):
    ≤ 1 ppm
  • Trace Impurities - Copper (Cu):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 1 ppm
  • Trace Impurities - Lead (Pb):
    ≤ 5 ppm

Specification Test Results

Related Information