Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Urea
Urea Ph. Eur., Macron Fine Chemicals™
Urea Ph. Eur., Macron Fine Chemicals™
Catalog # P123-38
Supplier:
MACRON AVANTOR BRAND
CAS Number:
SDS
Expand image
Urea Ph. Eur., Macron Fine Chemicals™
Catalog # P123-38
Supplier:
MACRON AVANTOR BRAND
CAS Number:
SDS
Specifications
Pk:
20 kg
Seq:
4241
Specifications
About this item
Product Details
Documentation
:
:
60.06 g/mol
CAS Number:
57-13-6
Formula:
NH₂CONH₂
MDL Number:
MFCD00008022
Product Details
Documentation