Order Entry
Germany
ContactUsLinkComponent
Sodium hydrogen carbonate Ph. Eur., Macron Fine Chemicals™
Sodium hydrogen carbonate Ph. Eur., Macron Fine Chemicals™
Catalog # P132-38
CAS Number:  
undefined
Sodium hydrogen carbonate Ph. Eur., Macron Fine Chemicals™
Catalog # P132-38
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :84.01 g/mol
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528