Order Entry
Germany
ContactUsLinkComponent
Potassium peroxodisulphate ≥99.0%, AR® ACS, Macron Fine Chemicals™
Oxidizing
Health
Potassium peroxodisulphate ≥99.0%, AR® ACS, Macron Fine Chemicals™
Catalog # 7076-20
CAS Number:  
undefined
Oxidizing
Health
Potassium peroxodisulphate ≥99.0%, AR® ACS, Macron Fine Chemicals™
Catalog # 7076-20
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (K₂S₂O₈):
    ≥99.0 %
  • ACS - Chlorine Compounds (as Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.0005 %
  • ACS - Manganese (Mn):
    ≤0.0002 %

Specification Test Results

About this item

 :
 :270.32 g/mol
Merck Index:13,07741
Melting Pt:100 °C
Density:2.477 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-781-8
UN:1492
ADR:5.1,III
 :GHS03
 :GHS08
CAS Number:7727-21-1
Formula:K₂S₂O₈
MDL Number:MFCD00011386