Order Entry
Germany
ContactUsLinkComponent
Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Oxidizing
Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Catalog # 6715-20
CAS Number:  
undefined
Oxidizing
Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Catalog # 6715-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 100.5 %
  • Assay (KNO₃):
    99.0 - 100.5 %
  • Solubility:
    Passes Test
  • Loss on Drying at 105°C:
    ≤1.0 %
  • Reaction:
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Identification for Sodium:
    Passes Test
  • Sodium (Na), For Information Only:
    Passes Test
  • Chlorate:
    Passes Test
  • Chloride (Cl):
    ≤0.058 %
  • Arsenic (As):
    ≤0.0003 %
  • Heavy Metals (as Pb):
    ≤0.0020 %
  • Lead (Pb):
    ≤0.0010 %

Specification Test Results

About this item

 :
 :101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
REACH:01-2119488224-35
 :GHS03
CAS Number:7757-79-1
Formula:KNO₃
MDL Number:MFCD00011409